missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-gamma-Aminobutyric acid, 98%, ACROS Organics™

Brand:  Acros Organics 376570050

Code : 29

Additional Details : CAS Number : 57294-38-9 Weight : 0.00500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10463103

  • £68.90 / 5g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

N-BOC-γ-Aminobutyric acid
97.5
97.5 to 102.5%
C9H17NO4
203.24
57294-38-9
100.0
98%
(CH3)3COC(=O)NH(CH2)3CO2H
MFCD00037313

Specifications

N-BOC-γ-Aminobutyric acid
97.5
Authentic
Powder
98%
54° to 60°C
203.24
5g
57294-38-9
100.0
Glass bottle
97.5 to 102.5%
C9H17NO4
(CH3)3COC(=O)NH(CH2)3CO2H
MFCD00037313
White to off-white
Videos
SDS