missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-DL-alpha-phenylglycinol, ≥98%

Brand:  Alfa Aesar™ H64489.06

Code : NEW

Additional Details : CAS Number : 67341-01-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15411729

  • £73.14 / 5g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-DL-alpha-phenylglycinol
237.3
4688247
67341-01-9
MFCD01883039
2-(Boc-amino)-2-phenylethanol; Boc-DL-Phg-ol

Specifications

N-Boc-DL-alpha-phenylglycinol
Powder
237.3
4688247
White to off-white
5g
67341-01-9
≥98%
MFCD01883039
2-(Boc-amino)-2-phenylethanol; Boc-DL-Phg-ol
141° to 144°C
Videos
SDS