missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-Diethanolamine, 98%, ACROS Organics™

Brand:  Acros Organics 440750250

Code : 29

Additional Details : CAS Number : 103898-11-9 Weight : 0.07700kg


Quantity 25mL

 View more versions of this product

Product Code. 11965501

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

N-BOC-Diethanolamine
97.5
≥97.5%
C9H19NO4
tert-Butyl N, N-bis(2-hydroxyethyl)carbamate
103898-11-9
100.0
98%
205.26

Specifications

N-BOC-Diethanolamine
97.5
Authentic
≥97.5%
1.085g/mL
205.26
Colorless
103898-11-9
100.0
Liquid or Oil
98%
C9H19NO4
25mL
tert-Butyl N, N-bis(2-hydroxyethyl)carbamate
Videos
SDS