missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-D-valinol, 95%

Brand:  Alfa Aesar™ H61149.06

Code : NEW

Additional Details : CAS Number : 106391-87-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15417658

  • £76.90 / 5g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-D-valinol
203.28
N-(tert-Butoxycarbonyl)-D-valinol
106391-87-1
MFCD00235960

Specifications

N-Boc-D-valinol
Liquid
203.28
1.4510
0.995g/mL
Pale yellow
106391-87-1
95%
MFCD00235960
N-(tert-Butoxycarbonyl)-D-valinol
218°C
5g
Videos
SDS