missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-D-styrylalanine dicyclohexylammonium salt, 98%

Brand:  Alfa Aesar™ H62782.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15457836

  • £99.64 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-D-styrylalanine dicyclohexylammonium salt
261380-19-2
472.66
MFCD01860890

Specifications

N-Boc-D-styrylalanine dicyclohexylammonium salt
Powder
472.66
White
261380-19-2
98%
MFCD01860890
1g
Videos
SDS