missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-D-phenylalanine N-succinimidyl ester, 97%

Brand:  Alfa Aesar™ H62763.06

Code : NEW

Additional Details : CAS Number : 3674-18-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15439248

  • £72.50 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-D-phenylalanine N-succinimidyl ester
362.38
1555676
3674-18-8
MFCD00069688
Boc-D-Phe-Osu; (R)-2,5-dioxopyrrolidin-1-yl 2-(Boc-amino)-3-phenylpropionate

Specifications

N-Boc-D-phenylalanine N-succinimidyl ester
Powder
362.38
1555676
White
5g
3674-18-8
97%
MFCD00069688
Boc-D-Phe-Osu; (R)-2,5-dioxopyrrolidin-1-yl 2-(Boc-amino)-3-phenylpropionate
149° to 153°C
Moisture Sensitive
Videos
SDS