missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-D-Phenylalaninal, 97%, Thermo Scientific™

Catalog Number p-3523510
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
77119-85-8
C14H19NO3
249.31
MFCD00274187
ZJTYRNPLVNMVPQ-GFCCVEGCSA-N
n-boc-d-phenylalaninal, boc-d-phenylalaninal, r-+-2-tert-butoxycarbonylamino-3-phenylpropanal, r-tert-butyl 1-oxo-3-phenylpropan-2-yl carbamate, n-boc-2 r-3-phenylpropanal, tert-butyl n-2r-1-oxo-3-phenylpropan-2-yl carbamate, tert-butyl 2r-1-oxo-3-phenylpropan-2-yl carbamate, carbamic acid, 1r-1-formyl-2-phenylethyl-, 1,1-dimethylethyl ester, boc-d-phe-h, pubchem11217
7020444
tert-butyl N-[(2R)-1-oxo-3-phenylpropan-2-yl]carbamate
CC(C)(C)OC(=O)N[C@H](CC1=CC=CC=C1)C=O
This item is not returnable. View return policy
77119-85-8
C14H19NO3
249.31
MFCD00274187
ZJTYRNPLVNMVPQ-GFCCVEGCSA-N
n-boc-d-phenylalaninal, boc-d-phenylalaninal, r-+-2-tert-butoxycarbonylamino-3-phenylpropanal, r-tert-butyl 1-oxo-3-phenylpropan-2-yl carbamate, n-boc-2 r-3-phenylpropanal, tert-butyl n-2r-1-oxo-3-phenylpropan-2-yl carbamate, tert-butyl 2r-1-oxo-3-phenylpropan-2-yl carbamate, carbamic acid, 1r-1-formyl-2-phenylethyl-, 1,1-dimethylethyl ester, boc-d-phe-h, pubchem11217
7020444
tert-butyl N-[(2R)-1-oxo-3-phenylpropan-2-yl]carbamate
CC(C)(C)OC(=O)N[C@H](CC1=CC=CC=C1)C=O
Melting Point 85.0°C to 89.0°C
CAS Min % 96.0
CAS Max % 100.0
Color Tan or White
Assay Percent Range 96% min. (GC)
Infrared Spectrum Authentic
Specific Rotation + 45.00
Formula Weight 249.31
Percent Purity 97%
Quantity 250 mg
Physical Form Crystalline Powder
Chemical Name or Material N-BOC-D-Phenylalaninal
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'