missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-leucinol, 98%, Thermo Scientific Chemicals

Catalog Number 15499018
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 106930-51-2
Boiling Point 213°C
Color Colorless
Molecular Formula C11H23NO3
MDL Number MFCD00235931
Synonym boc-d-leucinol, n-boc-d-leucinol, r-tert-butyl 1-hydroxy-4-methylpentan-2-yl carbamate, tert-butyl n-2r-1-hydroxy-4-methylpentan-2-yl carbamate, r-n-tert-butoxycarbonyl leucinol, ksc498g1f, r-1-hydroxymethyl-3-methyl-butyl-carbamic acid tert-butyl ester, 2 r-t-butoxycarbonylamino-4-methylpentanol, tert-butyl r-1-hydroxy-4-methylpentan-2-yl carbamate
Refractive Index 1.449
InChI Key LQTMEOSBXTVYRM-SECBINFHSA-N
SMILES CC(C)CC(CO)NC(=O)OC(C)(C)C
IUPAC Name tert-butyl N-[(2R)-1-hydroxy-4-methylpentan-2-yl]carbamate
Molecular Weight (g/mol) 217.309
PubChem CID 16211316
Formula Weight 217.31
Percent Purity 98%
Density 0.983 g/mL
Quantity 1 g
Flash Point 110°C (230°F)
Physical Form Liquid
Chemical Name or Material N-Boc-D-leucinol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'