missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-D-glucosamine, 97%

Brand:  Alfa Aesar™ H55682.03

Code : NEW

Additional Details : CAS Number : 75251-80-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15461998

  • £10.60 / 1g


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-D-glucosamine
279.29
3555040
75251-80-8
MFCD00134540

Specifications

N-Boc-D-glucosamine
Powder
279.29
MFCD00134540
White
75251-80-8
97%
194°C (dec.)
3555040
1g
Videos
SDS