missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-D-aspartic acid 4-tert-butyl ester dicyclohexylammonium salt, 98%

Brand:  Alfa Aesar™ H62664.03

Code : NEW

Additional Details : CAS Number : 200334-95-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15497696

  • £59.78 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-D-aspartic acid 4-tert-butyl ester dicyclohexylammonium salt
470.64
Boc-D-Asp(OtBu)-OH.DCHA; Dicyclohexylammonium (R)-2-(Boc-amino)-4-(tert-butoxy)-4-oxobutyrate
200334-95-8
MFCD00076911

Specifications

N-Boc-D-aspartic acid 4-tert-butyl ester dicyclohexylammonium salt
Solid
470.64
Boc-D-Asp(OtBu)-OH.DCHA; Dicyclohexylammonium (R)-2-(Boc-amino)-4-(tert-butoxy)-4-oxobutyrate
200334-95-8
98%
MFCD00076911
1g
Videos
SDS