missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-aspartic acid 1-tert-butyl ester, 98%, Thermo Scientific Chemicals

Catalog Number 15462809
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
77004-75-2
C13H23NO6
289.328
MFCD00236827
RAUQRYTYJIYLTF-MRVPVSSYSA-N
boc-d-asp-otbu, r-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-boc-d-aspartic acid 1-tert-butyl ester, r-4-tert-butoxy-3-boc-amino-4-oxobutanoic acid, 3r-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-d-aspotbu, n-boc-d-asparticacid1-tert-butyl ester, r-4-tert-butoxy-3-boc-amino-4-oxobutanoicacid, n-tert-butyloxycarbonyl-d-aspartic acid 1-tert-butyl ester
7019660
(3R)-4-[(2-methylpropan-2-yl)oxy]-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OC(C)(C)C
This item is not returnable. View return policy
77004-75-2
C13H23NO6
289.328
MFCD00236827
RAUQRYTYJIYLTF-MRVPVSSYSA-N
boc-d-asp-otbu, r-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-boc-d-aspartic acid 1-tert-butyl ester, r-4-tert-butoxy-3-boc-amino-4-oxobutanoic acid, 3r-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-d-aspotbu, n-boc-d-asparticacid1-tert-butyl ester, r-4-tert-butoxy-3-boc-amino-4-oxobutanoicacid, n-tert-butyloxycarbonyl-d-aspartic acid 1-tert-butyl ester
7019660
(3R)-4-[(2-methylpropan-2-yl)oxy]-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Beilstein 3553120
Formula Weight 289.33
Percent Purity 98%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material N-Boc-D-aspartic acid 1-tert-butyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'