missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-aniline, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Melting Point | 133°C to 137°C |
Color | Brown to White |
Linear Formula | C6H5NHCO2C(CH3)3 |
Quantity | 5 g |
Synonym | n-boc-aniline, tert-butyl phenylcarbamate, n-boc aniline, t-butyl carbanilate, tert-butyl carbanilate, carbanilic acid, tert-butyl ester, carbamic acid, phenyl-, 1,1-dimethylethyl ester, n-tert-butoxycarbonyl aniline, tert-butyl n-phenylurethane, carbanilic acid tert-butyl ester |
IUPAC Name | tert-butyl N-phenylcarbamate |
PubChem CID | 137930 |
Formula Weight | 193.25 |
Percent Purity | 0.97 |
Physical Form | Solid |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'