missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-4-oxo-L-proline methyl ester, 97%, ACROS Organics™

Brand:  Acros Organics 449270050

Code : NEW

Additional Details : CAS Number : 102195-80-2 Weight : 0.01000kg


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 13458648

  • £152.11 / 5g

Estimated Shipment
12-10-2020

Please to purchase this item. Need a web account? Register with us today!



Specifications

Chemical Identifiers

102195-80-2
243.26
boc-4-oxo-pro-ome, n-boc-4-oxo-l-proline methyl ester, s-1-tert-butyl 2-methyl 4-oxopyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2s-4-oxopyrrolidine-1,2-dicarboxylate, 2s-1-boc-4-oxo-proline methyl ester, methyl 1-boc-4-oxo-proline, 1-boc-4-oxo-l-proline methyl ester, tert-butyl 2s-2-methoxycarbonyl-4-oxopyrrolidine-1-carboxylate, 2s-4-oxo-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid methyl ester, 1,2-pyrrolidinedicarboxylic acid, 4-oxo-, 1-1,1-dimethylethyl 2-methyl ester, 2s
1-O-tert-butyl 2-O-methyl (2S)-4-oxopyrrolidine-1,2-dicarboxylate
C11H17NO5
UPBHYYJZVWZCOZ-QMMMGPOBSA-N
10988485
CC(C)(C)OC(=O)N1CC(=O)CC1C(=O)OC

Specifications

N-BOC-4-oxo-L-proline methyl ester
Glass bottle
5g
96% min. (GC)
boc-4-oxo-pro-ome, n-boc-4-oxo-l-proline methyl ester, s-1-tert-butyl 2-methyl 4-oxopyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2s-4-oxopyrrolidine-1,2-dicarboxylate, 2s-1-boc-4-oxo-proline methyl ester, methyl 1-boc-4-oxo-proline, 1-boc-4-oxo-l-proline methyl ester, tert-butyl 2s-2-methoxycarbonyl-4-oxopyrrolidine-1-carboxylate, 2s-4-oxo-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid methyl ester, 1,2-pyrrolidinedicarboxylic acid, 4-oxo-, 1-1,1-dimethylethyl 2-methyl ester, 2s
CC(C)(C)OC(=O)N1CC(=O)CC1C(=O)OC
243.26
243.26
>110°C
42°C to 45°C
102195-80-2
C11H17NO5
UPBHYYJZVWZCOZ-QMMMGPOBSA-N
1-O-tert-butyl 2-O-methyl (2S)-4-oxopyrrolidine-1,2-dicarboxylate
10988485
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • N-BOC-4-oxo-L-proline methyl ester

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documents