missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-4-oxo-L-proline, 98%, Thermo Scientific Chemicals

Catalog Number p-4813571
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
84348-37-8
C10H15NO5
229.23
MFCD01860669
CKYGSXRXTIKGAJ-UHFFFAOYNA-N
n-boc-4-oxo-l-proline, s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, n-t-boc-4-oxo-l-proline, boc-4-oxo-pro-oh, n-tert-boc-4-oxo-l-proline, 2s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, 1-tert-butoxycarbonyl-4-oxoproline, 2s-1-tert-butoxycarbonyl-4-oxo-pyrrolidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl-4-oxopyrrolidine-2-carboxylic acid, s-4-oxo-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
11593804
(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]-4-oxopyrrolidine-2-carboxylic acid
CC(C)(C)OC(=O)N1CC(=O)CC1C(O)=O
This item is not returnable. View return policy
84348-37-8
C10H15NO5
229.23
MFCD01860669
CKYGSXRXTIKGAJ-UHFFFAOYNA-N
n-boc-4-oxo-l-proline, s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, n-t-boc-4-oxo-l-proline, boc-4-oxo-pro-oh, n-tert-boc-4-oxo-l-proline, 2s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, 1-tert-butoxycarbonyl-4-oxoproline, 2s-1-tert-butoxycarbonyl-4-oxo-pyrrolidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl-4-oxopyrrolidine-2-carboxylic acid, s-4-oxo-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
11593804
(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]-4-oxopyrrolidine-2-carboxylic acid
CC(C)(C)OC(=O)N1CC(=O)CC1C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 155°C to 163°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Formula Weight 229.23
Percent Purity 98%
Chemical Name or Material N-BOC-4-oxo-L-proline
compliance-icons
Product Identifier
  • N-BOC-4-oxo-L-proline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'