missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-4-Hydroxypiperidine, 97%, ACROS Organics™

Brand:  Acros Organics 368270250

Code : 29

Additional Details : CAS Number : 109384-19-2 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10794943

  • £78.50 / 25g


EU Stock 3
Add to basket


Specifications

Chemical Identifiers

N-BOC-4-Hydroxypiperidine
109384-19-2
100.0
97%
201.27
N-(tert-Butoxycarbonyl)-4-hydroxypiperidine
97%
96.0
97%
C10H19NO3
MFCD01075174

Specifications

N-BOC-4-Hydroxypiperidine
109384-19-2
100.0
Glass bottle
97%
C10H19NO3
201.27
25g
N-(tert-Butoxycarbonyl)-4-hydroxypiperidine
97%
96.0
Authentic
Powder
97%
60.0° to 65.0°C
MFCD01075174
White to cream
Videos
SDS