missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-4-fluoro-L-phenylalanine, 95%

Brand:  Alfa Aesar™ H61370.03

Code : NEW

Additional Details : CAS Number : 41153-30-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15447898

  • £73.10 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-4-fluoro-L-phenylalanine
283.3
4323475
41153-30-4
MFCD00079672

Specifications

N-Boc-4-fluoro-L-phenylalanine
Powder
283.3
4323475
83° to 86°C
41153-30-4
95%
MFCD00079672
White
1g
Videos
SDS