missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-3-trifluoromethyl-D-phenylalanine, 95%

Brand:  Alfa Aesar™ H61496.06

Code : NEW

Additional Details : CAS Number : 82317-82-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15488048

  • £421.88 / 5g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-3-trifluoromethyl-D-phenylalanine
333.3
5133890
82317-82-6
MFCD00672525
Boc-D-3-trifluoromethylphe

Specifications

N-Boc-3-trifluoromethyl-D-phenylalanine
Solid
333.3
5133890
135° to 138°C
82317-82-6
95%
MFCD00672525
Boc-D-3-trifluoromethylphe
5g
Videos
SDS