missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-3-piperidone, 97%, Thermo Scientific Chemicals

Catalog Number p-3777577
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
98977-36-7
C10H17NO3
199.25
MFCD01631193
RIFXIGDBUBXKEI-UHFFFAOYSA-N
1-boc-3-piperidone, 1-boc-3-piperidinone, n-boc-3-piperidone, 1-n-boc-3-piperidone, n-boc-3-piperidinone, boc-3-piperidone, 1-tert-butoxycarbonyl-3-piperidone, 3-oxo-piperidine-1-carboxylic acid tert-butyl ester, 3-oxopiperidine, n-boc protected, tert-butyl 3-oxopiperidinecarboxylate
2756825
tert-butyl 3-oxopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(=O)C1
This item is not returnable. View return policy
98977-36-7
C10H17NO3
199.25
MFCD01631193
RIFXIGDBUBXKEI-UHFFFAOYSA-N
1-boc-3-piperidone, 1-boc-3-piperidinone, n-boc-3-piperidone, 1-n-boc-3-piperidone, n-boc-3-piperidinone, boc-3-piperidone, 1-tert-butoxycarbonyl-3-piperidone, 3-oxo-piperidine-1-carboxylic acid tert-butyl ester, 3-oxopiperidine, n-boc protected, tert-butyl 3-oxopiperidinecarboxylate
2756825
tert-butyl 3-oxopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(=O)C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 37°C to 39°C
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 199.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-BOC-3-piperidone
Show More Show Less
compliance-icons
Product Identifier
  • N-BOC-3-piperidone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'