missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-methyl-L-phenylalanine, 98%, Thermo Scientific Chemicals

Catalog Number 15409318
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
114873-06-2
C15H21NO4
279.336
MFCD01862944
HBBXWMALJNZDOM-LBPRGKRZSA-N
boc-phe 3-me-oh, boc-3-methyl-l-phenylalanine, boc-l-3-methylphenylalanine, boc-l-3-methylphe, boc-3-methy-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-m-tolyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-3-methylphenyl propanoic acid, boc-l-3-methyl phenylalanine, boc-phe 3-me-oh tlc, 3-methyl-n-t-butoxycarbonyl-l-phenylalanine
2761470
(2S)-3-(3-methylphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC1=CC(=CC=C1)CC(C(=O)O)NC(=O)OC(C)(C)C
This item is not returnable. View return policy
114873-06-2
C15H21NO4
279.336
MFCD01862944
HBBXWMALJNZDOM-LBPRGKRZSA-N
boc-phe 3-me-oh, boc-3-methyl-l-phenylalanine, boc-l-3-methylphenylalanine, boc-l-3-methylphe, boc-3-methy-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-m-tolyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-3-methylphenyl propanoic acid, boc-l-3-methyl phenylalanine, boc-phe 3-me-oh tlc, 3-methyl-n-t-butoxycarbonyl-l-phenylalanine
2761470
(2S)-3-(3-methylphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC1=CC(=CC=C1)CC(C(=O)O)NC(=O)OC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Beilstein 8989098
Formula Weight 279.33
Percent Purity 98%
Quantity 1 g
Physical Form Solid
Chemical Name or Material N-Boc-3-methyl-L-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'