missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-3-methyl-L-phenylalanine, 98%

Brand:  Alfa Aesar™ H62860.06

Code : NEW

Additional Details : CAS Number : 114873-06-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15419318

  • £204.60 / 5g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-3-methyl-L-phenylalanine
279.33
8989098
114873-06-2
MFCD01862944
Boc-L-3-methylphe; Boc-Phe(3-Me)-OH

Specifications

N-Boc-3-methyl-L-phenylalanine
Solid
279.33
8989098
5g
114873-06-2
98%
MFCD01862944
Boc-L-3-methylphe; Boc-Phe(3-Me)-OH
Videos
SDS