missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-3-bromo-D-phenylalanine, 98%

Brand:  Alfa Aesar™ H62761.03

Code : NEW

Additional Details : CAS Number : 261360-77-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15479238

  • £104.90 / 1g


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-3-bromo-D-phenylalanine
344.2
(R)-N-Boc-3-bromophenylalanine; Boc-Phe(3-Br)-OH
261360-77-4
MFCD01317704

Specifications

N-Boc-3-bromo-D-phenylalanine
Powder
344.2
(R)-N-Boc-3-bromophenylalanine; Boc-Phe(3-Br)-OH
1g
261360-77-4
98%
MFCD01317704
White
Videos
SDS