missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-(4-pyridyl)-D-alanine, 95%, Thermo Scientific Chemicals

Catalog Number 15428768
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
37535-58-3
C13H18N2O4
266.297
MFCD00672529
FNYWDMKESUACOU-SNVBAGLBSA-N
boc-d-4-pyridylalanine, boc-d-4-pal-oh, r-2-tert-butoxycarbonyl amino-3-pyridin-4-yl propanoic acid, boc-3-4-pyridyl-d-alanine, boc-d-4-pyridylala, boc-4'-pyridyl-d-ala, boc-3-4-pyridyl-d-ala-oh, n-tert-butoxycarbonyl-4-pyridyl-d-alanine, boc-d-3-4-pyridyl-alanine, r-2-tert-butoxycarbonylamino-3-pyridin-4-yl-propionic acid
2734503
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-4-ylpropanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=NC=C1)C(=O)O
This item is not returnable. View return policy
37535-58-3
C13H18N2O4
266.297
MFCD00672529
FNYWDMKESUACOU-SNVBAGLBSA-N
boc-d-4-pyridylalanine, boc-d-4-pal-oh, r-2-tert-butoxycarbonyl amino-3-pyridin-4-yl propanoic acid, boc-3-4-pyridyl-d-alanine, boc-d-4-pyridylala, boc-4'-pyridyl-d-ala, boc-3-4-pyridyl-d-ala-oh, n-tert-butoxycarbonyl-4-pyridyl-d-alanine, boc-d-3-4-pyridyl-alanine, r-2-tert-butoxycarbonylamino-3-pyridin-4-yl-propionic acid
2734503
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-4-ylpropanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=NC=C1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 223°C to 229°C
Color White
Beilstein 4195478
Formula Weight 266.29
Percent Purity 95%
Quantity 250 mg
Sensitivity Air and moisture sensitive
Physical Form Powder
Chemical Name or Material N-Boc-3-(4-pyridyl)-D-alanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'