missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-3-(4-Cyanophenyl)oxaziridine, 98+%, Thermo Scientific Chemicals

Catalog Number p-71075
Click to view available options
Quantity:
500 mg
2 g
Packaging:
Glass bottle
This item is not returnable. View return policy
150884-56-3
C13H14N2O3
246.27
MFCD00800493
ACXPNVRTMHEHMQ-UHFFFAOYSA-N
n-boc-3-4-cyanophenyl oxaziridine, tert-butyl 3-4-cyanophenyl-1,2-oxaziridine-2-carboxylate, tert-butyl 3-4-cyanophenyl oxaziridine-2-carboxylate, 2-oxaziridinecarboxylicacid, 3-4-cyanophenyl-, 1,1-dimethylethyl ester, 2r,3s-rel-9ci, acmc-20n62r, n-boc-3-4-cyanophenyl oxaziridine, 98+%, 2-tert-butoxycarbonyl-3-4-cyanophenyl oxaziridine, n-tert-butoxycarbonyl-3-4-cyanophenyl-oxaziridine, 3-4-cyano-phenyl-oxaziridine-2-carboxylic acid tert-butyl ester, 3-4-cyano-phenyl oxaziridine-2-carboxylic acid tert-butyl ester
2725038
tert-butyl 3-(4-cyanophenyl)oxaziridine-2-carboxylate
CC(C)(C)OC(=O)N1C(O1)C2=CC=C(C=C2)C#N
This item is not returnable. View return policy
150884-56-3
C13H14N2O3
246.27
MFCD00800493
ACXPNVRTMHEHMQ-UHFFFAOYSA-N
n-boc-3-4-cyanophenyl oxaziridine, tert-butyl 3-4-cyanophenyl-1,2-oxaziridine-2-carboxylate, tert-butyl 3-4-cyanophenyl oxaziridine-2-carboxylate, 2-oxaziridinecarboxylicacid, 3-4-cyanophenyl-, 1,1-dimethylethyl ester, 2r,3s-rel-9ci, acmc-20n62r, n-boc-3-4-cyanophenyl oxaziridine, 98+%, 2-tert-butoxycarbonyl-3-4-cyanophenyl oxaziridine, n-tert-butoxycarbonyl-3-4-cyanophenyl-oxaziridine, 3-4-cyano-phenyl-oxaziridine-2-carboxylic acid tert-butyl ester, 3-4-cyano-phenyl oxaziridine-2-carboxylic acid tert-butyl ester
2725038
tert-butyl 3-(4-cyanophenyl)oxaziridine-2-carboxylate
CC(C)(C)OC(=O)N1C(O1)C2=CC=C(C=C2)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 57.0°C to 63.0°C
Color Beige to White
Elemental Analysis 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 500 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in ether,methylene chloride,chloroform,,etc
Formula Weight 246.27
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material N-BOC-3-(4-Cyanophenyl)oxaziridine, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • N-BOC-3-(4-Cyanophenyl)oxaziridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'