missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-3-(3-pyridyl)-L-alanine, 98%

Brand:  Alfa Aesar™ H62063.03

Code : NEW

Additional Details : CAS Number : 117142-26-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15478648

  • £119.78 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-3-(3-pyridyl)-L-alanine
266.29
7625087
117142-26-4
MFCD00079681
Boc-3-(3-pyridyl)-Ala-OH; (S)-2-(Boc-amino)-3-(3-pyridyl)propionic acid

Specifications

N-Boc-3-(3-pyridyl)-L-alanine
Solid
266.29
7625087
136° to 142°C
117142-26-4
98%
MFCD00079681
Boc-3-(3-pyridyl)-Ala-OH; (S)-2-(Boc-amino)-3-(3-pyridyl)propionic acid
1g
Videos
SDS