missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-3-(2-furyl)-D-alanine dicyclohexylammonium salt, 98%

Brand:  Alfa Aesar™ H62489.03

Code : NEW

Additional Details : CAS Number : 331730-09-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15477486

  • £183.38 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-3-(2-furyl)-D-alanine dicyclohexylammonium salt
436.58
Boc-β-(2-furyl)-D-Ala-OH.DCHA
331730-09-7
MFCD01860889

Specifications

N-Boc-3-(2-furyl)-D-alanine dicyclohexylammonium salt
Powder
436.58
Boc-β-(2-furyl)-D-Ala-OH.DCHA
1g
331730-09-7
98%
MFCD01860889
White
Videos
SDS