missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-2-cyclohexyl-L-glycine, 97%

Brand:  Alfa Aesar™ H62466.06

Code : NEW

Additional Details : CAS Number : 109183-71-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15408988

  • £113.20 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Description

Description

Applications
N-Boc-2-cyclohexyl-L-glycine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

N-Boc-2-cyclohexyl-L-glycine
257.33
5553687
109183-71-3
MFCD00133638
Boc-Chg-OH

Specifications

N-Boc-2-cyclohexyl-L-glycine
Solid
257.33
5553687
5g
109183-71-3
97%
MFCD00133638
Boc-Chg-OH
Videos
SDS