missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-2-chloro-L-phenylalanine, 95%

Brand:  Alfa Aesar™ H62756.03

Code : NEW

Additional Details : CAS Number : 114873-02-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409238

  • £52.15 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-2-chloro-L-phenylalanine
299.75
8778440
114873-02-8
MFCD00672514
Boc-L-2-chlorophe; Boc-Phe(2-Cl)-OH

Specifications

N-Boc-2-chloro-L-phenylalanine
Solid
299.75
8778440
94° to 100°C
114873-02-8
95%
MFCD00672514
Boc-L-2-chlorophe; Boc-Phe(2-Cl)-OH
1g
Videos
SDS