missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-chloro-D-phenylalanine, 95%, Thermo Scientific™

Catalog Number p-7053573
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
80102-23-4
C14H18ClNO4
299.751
MFCD00801056
TUUQJNHCVFJMPU-LLVKDONJSA-N
boc-2-chloro-d-phenylalanine, boc-d-2-chlorophe, r-2-tert-butoxycarbonyl amino-3-2-chlorophenyl propanoic acid, boc-d-phe 2-cl-oh, boc-d-2-chlorophenylalanine, boc-d-2-chlooro phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-2-chlorophenyl propanoic acid, boc-l-phe 2-cl-oh, d-phenylalanine, 2-chloro-n-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxy carbonylamino-3-2-chlorophenyl propanoic acid
7021188
(2R)-3-(2-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1Cl)C(=O)O
This item is not returnable. View return policy
80102-23-4
C14H18ClNO4
299.751
MFCD00801056
TUUQJNHCVFJMPU-LLVKDONJSA-N
boc-2-chloro-d-phenylalanine, boc-d-2-chlorophe, r-2-tert-butoxycarbonyl amino-3-2-chlorophenyl propanoic acid, boc-d-phe 2-cl-oh, boc-d-2-chlorophenylalanine, boc-d-2-chlooro phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-2-chlorophenyl propanoic acid, boc-l-phe 2-cl-oh, d-phenylalanine, 2-chloro-n-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxy carbonylamino-3-2-chlorophenyl propanoic acid
7021188
(2R)-3-(2-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1Cl)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 94°C to 98°C
Beilstein 8422927
Formula Weight 299.75
Percent Purity 95%
Quantity 1 g
Physical Form Solid
Chemical Name or Material N-Boc-2-chloro-D-phenylalanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'