missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-2-chloro-D-phenylalanine, 95%

Brand:  Alfa Aesar™ H61107.06

Code : NEW

Additional Details : CAS Number : 80102-23-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15447608

  • £146.50 / 5g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-2-chloro-D-phenylalanine
299.75
8422927
80102-23-4
MFCD00801056
Boc-D-Phe(2-Cl)-OH; Boc-D-2-chlorophe

Specifications

N-Boc-2-chloro-D-phenylalanine
Solid
299.75
8422927
94° to 98°C
80102-23-4
95%
MFCD00801056
Boc-D-Phe(2-Cl)-OH; Boc-D-2-chlorophe
5g
Videos
SDS