Alfa Aesar™ N-Boc-2,4-dichloro-D-phenylalanine, 98%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H62706.MD

UNSPSC: 12352209

Code: NEW

Additional Details:
Additional Details: CAS Number: 114873-12-0 Weight: 0.00025kg


Quantity 250mg

Product Code. 15479188

Quantity Price
1 £ 24.38 / 250mg
Estimated Shipment date
from Supplier 09-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material N-Boc-2,4-dichloro-D-phenylalanine
CAS 114873-12-0
Formula Weight 334.2
MDL Number MFCD01862941
Synonym Boc-D-Phe(2,4-Cl2)-OH

Specification

Formula Weight 334.2
Melting Point 133° to 137°C
Physical Form Solid
Assay 98%
Chemical Name or Material N-Boc-2,4-dichloro-D-phenylalanine
MDL Number MFCD01862941
CAS 114873-12-0
Synonym Boc-D-Phe(2,4-Cl2)-OH
Quantity 250mg
Documentation

Technical documents