missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-1-(p-toluenesulfonyl)-L-histidine dicyclohexylammonium salt, 98%

Brand:  Alfa Aesar™ H62205.14

Code : NEW

Additional Details : CAS Number : 65057-34-3 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15477156

  • £75.40 / 25g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-1-(p-toluenesulfonyl)-L-histidine dicyclohexylammonium salt
590.77
Boc-His(Tos)-OH.DCHA; N-Boc-1-(p-tosyl)-L-histidine dicyclohexylammonium salt
65057-34-3
MFCD00039005

Specifications

N-Boc-1-(p-toluenesulfonyl)-L-histidine dicyclohexylammonium salt
Solid
590.77
Boc-His(Tos)-OH.DCHA; N-Boc-1-(p-tosyl)-L-histidine dicyclohexylammonium salt
65057-34-3
98%
MFCD00039005
25g
Videos
SDS