missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-1-(p-toluenesulfonyl)-D-histidine, 95%

Brand:  Alfa Aesar™ H63855.03

Code : NEW

Additional Details : CAS Number : 69541-68-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15480779

  • £28.73 / 1g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-1-(p-toluenesulfonyl)-D-histidine
409.46
Boc-D-His(Tos)-OH
69541-68-0
MFCD00037890

Specifications

N-Boc-1-(p-toluenesulfonyl)-D-histidine
Powder
409.46
Boc-D-His(Tos)-OH
1g
69541-68-0
95%
MFCD00037890
Pale yellow
Videos
SDS