missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-1-(p-toluenesulfonyl)-D-histidine, 95%

Brand:  Alfa Aesar™ H63855.06

Code : NEW

Additional Details : CAS Number : 69541-68-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15490779

  • £143.10 / 5g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-1-(p-toluenesulfonyl)-D-histidine
409.46
Boc-D-His(Tos)-OH
69541-68-0
MFCD00037890

Specifications

N-Boc-1-(p-toluenesulfonyl)-D-histidine
Powder
409.46
Boc-D-His(Tos)-OH
5g
69541-68-0
95%
MFCD00037890
Pale yellow
Videos
SDS