missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-1,3-diaminopropane, 97%, Thermo Scientific Chemicals

Catalog Number 11955531
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy
75178-96-0
C8H19N2O2
175.25
MFCD00210021
POHWAQLZBIMPRN-UHFFFAOYSA-O
n-boc-1,3-propanediamine, n-boc-1,3-diaminopropane, tert-butyl n-3-aminopropyl carbamate, tert-butyl 3-aminopropyl carbamate, n-3-aminopropyl carbamic acid tert-butyl ester, tert-butyl 3-aminopropylcarbamate, 1-boc-amino-1,3-propanediamine, 3-amino-propyl-carbamic acid tert-butyl ester, n-t-boc-1,3-propanediamine, n-tert-butoxycarbonyl-1,3-diaminopropane
2735700
tert-butyl N-(3-aminopropyl)carbamate
CC(C)(C)OC(=O)NCCC[NH3+]
This item is not returnable. View return policy
75178-96-0
C8H19N2O2
175.25
MFCD00210021
POHWAQLZBIMPRN-UHFFFAOYSA-O
n-boc-1,3-propanediamine, n-boc-1,3-diaminopropane, tert-butyl n-3-aminopropyl carbamate, tert-butyl 3-aminopropyl carbamate, n-3-aminopropyl carbamic acid tert-butyl ester, tert-butyl 3-aminopropylcarbamate, 1-boc-amino-1,3-propanediamine, 3-amino-propyl-carbamic acid tert-butyl ester, n-t-boc-1,3-propanediamine, n-tert-butoxycarbonyl-1,3-diaminopropane
2735700
tert-butyl N-(3-aminopropyl)carbamate
CC(C)(C)OC(=O)NCCC[NH3+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 22.0°C
Boiling Point 94.0°C to 96.0°C (0.2 Torr)
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Assay Percent Range 96% min. (GC)
Infrared Spectrum Authentic
Linear Formula (CH3)3CHOC(O)NH(CH2)3NH2
Packaging Glass Bottle
Formula Weight 174.24
Percent Purity 97%
Density 0.9980g/mL
Quantity 5 mL
Specific Gravity 0.998
Physical Form Liquid
Chemical Name or Material N-BOC-1, 3-diaminopropane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'