missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1,2,5,6-tetrahydropyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog Number 15482699
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
286961-14-6
C16H28BNO4
309.21
MFCD03840345
VVDCRJGWILREQH-UHFFFAOYSA-N
n-boc-1,2,5,6-tetrahydropyridine-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5,6-dihydropyridine-1 2h-carboxylate, 1-n-boc-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine, n-boc-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, tert-butyl 5,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-1 2h-carboxylate, 3,6-dihydro-2h-pyridine-1-n-boc-4-boronic acid pinacol ester, n-tert-butoxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, 1-boc-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine, tert-butyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2,3,6-tetrahydropyridine-1-carboxylate
4642098
tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(=CC1)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
286961-14-6
C16H28BNO4
309.21
MFCD03840345
VVDCRJGWILREQH-UHFFFAOYSA-N
n-boc-1,2,5,6-tetrahydropyridine-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5,6-dihydropyridine-1 2h-carboxylate, 1-n-boc-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine, n-boc-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, tert-butyl 5,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-1 2h-carboxylate, 3,6-dihydro-2h-pyridine-1-n-boc-4-boronic acid pinacol ester, n-tert-butoxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, 1-boc-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine, tert-butyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2,3,6-tetrahydropyridine-1-carboxylate
4642098
tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(=CC1)B1OC(C)(C)C(C)(C)O1

Reagent used for suzuki-Miyaura cross-coupling using palladium phosphine catalyst, palladium-catalyzed ligand-controlled regioselective Suzuki coupling, wrenchnolol derivative optimized for gene activation in cells. Also used in Preparation of several enzymatic inhibitors and receptor ligands, orally active anaplastic lymphoma kinase inhibitors and oxazolecarboxamides as diacylglycerol acyltransferase-1 inhibitors for treatment of obesity and diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent used for suzuki-Miyaura cross-coupling using palladium phosphine catalyst, palladium-catalyzed ligand-controlled regioselective Suzuki coupling, wrenchnolol derivative optimized for gene activation in cells. Also used in Preparation of several enzymatic inhibitors and receptor ligands, orally active anaplastic lymphoma kinase inhibitors and oxazolecarboxamides as diacylglycerol acyltransferase-1 inhibitors for treatment of obesity and diabetes.

Solubility
Soluble in most organic solvents. Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Solubility Information Soluble in most organic solvents. Insoluble in water.
Formula Weight 327.23
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-1,2,5,6-tetrahydropyridine-4-boronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'