missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-1-(2,4-dinitrophenyl)-L-histidine, 99%

Brand:  Alfa Aesar™ H63421.06

Code : NEW

Additional Details : CAS Number : 25024-53-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15480099

  • £32.40 / 5g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Light Sensitive, store in dark. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

N-Boc-1-(2,4-dinitrophenyl)-L-histidine
421.36
771922
25024-53-7
MFCD00065966
Boc-His(Dnp)-OH

Specifications

N-Boc-1-(2,4-dinitrophenyl)-L-histidine
Solid
421.36
771922
5g
25024-53-7
99%
MFCD00065966
Boc-His(Dnp)-OH
Light Sensitive
Videos
SDS