missing translation for 'onlineSavingsMsg'
Learn More

N-(Benzyloxycarbonyloxy)succinimide, 99%, Thermo Scientific Chemicals

Catalog Number p-7081951
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
13139-17-8
C12H11NO5
249.222
MFCD00005513
MJSHDCCLFGOEIK-UHFFFAOYSA-N
n-benzyloxycarbonyloxy succinimide, cbz-osu, cbz-onsu, benzyl 2,5-dioxopyrrolidin-1-yl carbonate, benzyl succinimido carbonate, z-osu, n-carbobenzoxyoxysuccinimide, n-benzyloxycarbonyloxy-succinimide, cbzosu, carbobenzoxy succinimide
83172
benzyl (2,5-dioxopyrrolidin-1-yl) carbonate
C1CC(=O)N(C1=O)OC(=O)OCC2=CC=CC=C2
This item is not returnable. View return policy
13139-17-8
C12H11NO5
249.222
MFCD00005513
MJSHDCCLFGOEIK-UHFFFAOYSA-N
n-benzyloxycarbonyloxy succinimide, cbz-osu, cbz-onsu, benzyl 2,5-dioxopyrrolidin-1-yl carbonate, benzyl succinimido carbonate, z-osu, n-carbobenzoxyoxysuccinimide, n-benzyloxycarbonyloxy-succinimide, cbzosu, carbobenzoxy succinimide
83172
benzyl (2,5-dioxopyrrolidin-1-yl) carbonate
C1CC(=O)N(C1=O)OC(=O)OCC2=CC=CC=C2

N-(N-Benzyloxycarbonyloxy)succinimide is a reagent used in the preparation of peptide thioacids view peptide precursors. Also used in the preparation of amino glycoside derivatives for the function of topoisomerase for both human and bacterial inhibition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(N-Benzyloxycarbonyloxy)succinimide is a reagent used in the preparation of peptide thioacids view peptide precursors. Also used in the preparation of amino glycoside derivatives for the function of topoisomerase for both human and bacterial inhibition.

Solubility
Soluble in chloroform, methanol, acetone (0.1 g/mL).

Notes
Moisture sensitive. Storage temperature 2-8°C. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY
Melting Point 77°C to 82°C
Quantity 5 g
Beilstein 1387927
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform,methanol,acetone (0.1g/mL).
Formula Weight 249.23
Percent Purity 99%
Chemical Name or Material N- (Benzyloxycarbonyloxy)succinimide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'