missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonyl-L-valine N-succinimidyl ester, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
N-Benzyloxycarbonyl-L-valine N-succinimidyl ester, is an amino acid which is widely used in pharmaceutical and food industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Benzyloxycarbonyl-L-valine N-succinimidyl ester, is an amino acid which is widely used in pharmaceutical and food industry.
Solubility
Soluble in 1mmol in 2mL DCM.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
N-Benzyloxycarbonyl-L-valine N-succinimidyl ester, is an amino acid which is widely used in pharmaceutical and food industry.
Solubility
Soluble in 1mmol in 2mL DCM.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Melting Point | 114°C to 118°C |
Color | White |
Molecular Formula | C17H20N2O6 |
Beilstein | 1555287 |
Synonym | z-val-osu, n-cbz-l-valine succinimidyl ester, s-2,5-dioxopyrrolidin-1-yl 2-benzyloxy carbonyl amino-3-methylbutanoate, l-valine,n-phenylmethoxy carbonyl-, 2,5-dioxo-1-pyrrolidinyl ester, zvalosu, 2,5-dioxopyrrolidin-1-yl 2s-2-benzyloxy carbonyl amino-3-methylbutanoate, cbz-val-osu, ksc496o1j, benzyl s-1-2,5-dioxo-1-pyrrolidinyl oxy carbonyl-2-methylpropyl carbamate, n-cbz-valinyl oxy-succinimide |
Solubility Information | Soluble in 1mmol in 2mL DCM. |
InChI Key | MFAOBGXYLNLLJE-HNNXBMFYSA-N |
SMILES | CC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O |
IUPAC Name | (2,5-dioxopyrrolidin-1-yl) (2S)-3-methyl-2-(phenylmethoxycarbonylamino)butanoate |
Molecular Weight (g/mol) | 348.36 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'