missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-D-glutamic acid 1-methyl ester, 98%

Brand:  Alfa Aesar™ H63008.03

Code : NEW

Additional Details : CAS Number : 26566-11-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15408096

  • £27.56 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

N-Benzyloxycarbonyl-D-glutamic acid 1-methyl ester
295.29
Z-D-Glu-OMe
26566-11-0
MFCD00798654

Specifications

N-Benzyloxycarbonyl-D-glutamic acid 1-methyl ester
Solid
295.29
Z-D-Glu-OMe
26566-11-0
98%
MFCD00798654
1g
Videos
SDS