missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-D-aspartic acid 4-tert-butyl ester hydrate, 98%

Brand:  Alfa Aesar™ H63757.06

Code : NEW

Additional Details : CAS Number : 71449-08-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15418506

  • £60.00 / 5g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Benzyloxycarbonyl-D-aspartic acid 4-tert-butyl ester hydrate
141.36
Z-D-Asp(OtBu)-OH.H2O
71449-08-6
MFCD00153327

Specifications

N-Benzyloxycarbonyl-D-aspartic acid 4-tert-butyl ester hydrate
Solid
141.36
Z-D-Asp(OtBu)-OH.H2O
71449-08-6
98%
MFCD00153327
5g
Videos
SDS