missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-2-methylalanine, 98%, Thermo Scientific Chemicals

Catalog Number 15469628
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
15030-72-5
C12H15NO4
237.255
MFCD00004190
QKVCSJBBYNYZNM-UHFFFAOYSA-N
z-aib-oh, n-cbz-2-methylalanine, 2-benzyloxy carbonyl amino-2-methylpropanoic acid, n-cbz-2-amino-2-methylpropanoic acid, z-2-aminoisobutyric acid, z-2-methylalanine, n-carbobenzyloxy-2-methylalanine, 2-carbobenzoxyamino isobutyric acid, n-benzyloxy carbonyl-2-methylalanine, alanine, 2-methyl-n-phenylmethoxy carbonyl
294936
2-methyl-2-(phenylmethoxycarbonylamino)propanoic acid
CC(C)(C(=O)O)NC(=O)OCC1=CC=CC=C1
This item is not returnable. View return policy
15030-72-5
C12H15NO4
237.255
MFCD00004190
QKVCSJBBYNYZNM-UHFFFAOYSA-N
z-aib-oh, n-cbz-2-methylalanine, 2-benzyloxy carbonyl amino-2-methylpropanoic acid, n-cbz-2-amino-2-methylpropanoic acid, z-2-aminoisobutyric acid, z-2-methylalanine, n-carbobenzyloxy-2-methylalanine, 2-carbobenzoxyamino isobutyric acid, n-benzyloxy carbonyl-2-methylalanine, alanine, 2-methyl-n-phenylmethoxy carbonyl
294936
2-methyl-2-(phenylmethoxycarbonylamino)propanoic acid
CC(C)(C(=O)O)NC(=O)OCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 65°C to 69°C (decomposition)
Color White
Beilstein 2218009
Formula Weight 237.25
Percent Purity 98%
Quantity 1 g
Physical Form Powder
Chemical Name or Material N-Benzyloxycarbonyl-2-methylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'