missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-1-Boc-L-tryptophan dicyclohexylammonium salt, 98%

Brand:  Alfa Aesar™ H62015.03

Code : NEW

Additional Details : CAS Number : 218938-57-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15436926

  • £24.70 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Benzyloxycarbonyl-1-Boc-L-tryptophan dicyclohexylammonium salt
619.79
Z-Trp(Boc)-OH.DCHA
218938-57-9
MFCD01318754

Specifications

N-Benzyloxycarbonyl-1-Boc-L-tryptophan dicyclohexylammonium salt
Solid
619.79
Z-Trp(Boc)-OH.DCHA
218938-57-9
98%
MFCD01318754
1g
Videos
SDS