Learn More
N-Benzylnaltrindole hydrochloride, Tocris Bioscience™
Selective non-peptide δ2 antagonist
Brand:  Tocris (Bio-Techne) 075450
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13447963
  • £458.64 / 50mg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
Potent δ2-selective opioid receptor antagonist with a long duration of action in vivo.
Specifications
Chemical Identifiers
1206487-81-1
541.088
n-benzylnaltrindole hydrochloride
C1CC1CN2CCC34C5C6=C(CC3(C2CC7=C4C(=C(C=C7)O)O5)O)C8=CC=CC=C8N6CC9=CC=CC=C9.Cl
C33H33ClN2O3
KDEBSUYWJUBPCB-REBVNRAYSA-N
131845649
Specifications
50mg
1206487-81-1
n-benzylnaltrindole hydrochloride
C1CC1CN2CCC34C5C6=C(CC3(C2CC7=C4C(=C(C=C7)O)O5)O)C8=CC=CC=C8N6CC9=CC=CC=C9.Cl
131845649
>98%
N-Benzylnaltrindole hydrochloride
C33H33ClN2O3
KDEBSUYWJUBPCB-REBVNRAYSA-N
541.088
541.09