missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylidenemethylamine, 99%, Thermo Scientific Chemicals

Catalog Number p-66458
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
622-29-7
MFCD00008294
HXTGGPKOEKKUQO-UHFFFAOYSA-N
n-benzylidenemethylamine, n-benzylidenemethanamine, benzylidenemethylamine, n-methylbenzaldimine, methanamine, n-phenylmethylene, benzylidenemethanamine, n-methylbenzylideneamine, methylamine, n-benzylidene, n-phenylmethylene methanamine, benzylidene-methyl-amine
73954
N-methyl-1-phenylmethanimine
CN=CC1=CC=CC=C1
This item is not returnable. View return policy
622-29-7
MFCD00008294
HXTGGPKOEKKUQO-UHFFFAOYSA-N
n-benzylidenemethylamine, n-benzylidenemethanamine, benzylidenemethylamine, n-methylbenzaldimine, methanamine, n-phenylmethylene, benzylidenemethanamine, n-methylbenzylideneamine, methylamine, n-benzylidene, n-phenylmethylene methanamine, benzylidene-methyl-amine
73954
N-methyl-1-phenylmethanimine
CN=CC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color White
Density 96%g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H9N
Linear Formula C6H5CH=NCH3
Refractive Index 1.5500 to 1.5540 (20°C, 589nm)
Quantity 5 g
Beilstein 07, 213
Molecular Weight (g/mol) 119.167
Formula Weight 119.17
Percent Purity ≥98.5% (GC)
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material N-Benzylidenemethylamine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • N-Benzylidenemethylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'