missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzylglycine ethyl ester, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
N-Benzylglycine ethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Benzylglycine ethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
N-Benzylglycine ethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Boiling Point | 102°C to 104°C (1mmHg) |
Beilstein | 2104816 |
Refractive Index | 1.5053 |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 193.25 |
Percent Purity | 97% |
Density | 1.048 |
Quantity | 25 g |
Flash Point | 112°C (233°F) |
Chemical Name or Material | N-Benzylglycine ethyl ester |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'