missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylglycine ethyl ester, 96%, Thermo Scientific Chemicals

Catalog Number 10172940
70.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
6436-90-4
C11H15NO2
193.25
MFCD00009174
ULOLIZHBYWAICY-UHFFFAOYSA-N
n-benzylglycine ethyl ester, ethyl 2-benzylamino acetate, ethyl n-benzylglycinate, n-bzl-gly-oet, bzl-gly-oet, glycine, n-phenylmethyl-, ethyl ester, n-benzyl glycine ethyl ester, ethyl n-phenylmethyl glycinate, ethyl benzylamino acetate, n-benzyl-glycine ethyl ester
80908
ethyl 2-(benzylamino)acetate
CCOC(=O)CNCC1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
6436-90-4
C11H15NO2
193.25
MFCD00009174
ULOLIZHBYWAICY-UHFFFAOYSA-N
n-benzylglycine ethyl ester, ethyl 2-benzylamino acetate, ethyl n-benzylglycinate, n-bzl-gly-oet, bzl-gly-oet, glycine, n-phenylmethyl-, ethyl ester, n-benzyl glycine ethyl ester, ethyl n-phenylmethyl glycinate, ethyl benzylamino acetate, n-benzyl-glycine ethyl ester
80908
ethyl 2-(benzylamino)acetate
CCOC(=O)CNCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0300g/mL
Boiling Point 140°C to 142°C (10.0 mmHg)
Flash Point 112°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2NHCH2CO2C2H5
Refractive Index 1.5045 to 1.5065
Quantity 50 g
Beilstein 12, 1060
Specific Gravity 1.03
Formula Weight 193.24
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material N-Benzylglycine ethyl ester, 96%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'