missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylglycine, 98+%, Thermo Scientific Chemicals

Catalog Number p-7058434
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
17136-36-6
C9H11NO2
165.192
MFCD00190772
KGSVNOLLROCJQM-UHFFFAOYSA-N
n-benzylglycine, 2-benzylamino acetic acid, n-benzyl glycine, glycine, n-phenylmethyl, n-phenylmethyl glycine, benzylglycine, benzylamino acetic acid, n-benzylglycin, bzl-gly-oh
86965
2-(benzylamino)acetic acid
C1=CC=C(C=C1)CNCC(=O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
17136-36-6
C9H11NO2
165.192
MFCD00190772
KGSVNOLLROCJQM-UHFFFAOYSA-N
n-benzylglycine, 2-benzylamino acetic acid, n-benzyl glycine, glycine, n-phenylmethyl, n-phenylmethyl glycine, benzylglycine, benzylamino acetic acid, n-benzylglycin, bzl-gly-oh
86965
2-(benzylamino)acetic acid
C1=CC=C(C=C1)CNCC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∽210°C (decomposition)
Beilstein 2209788
Formula Weight 165.19
Percent Purity ≥98%
Quantity 5 g
Chemical Name or Material N-Benzylglycine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'