missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyldimethylamine, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081932
12.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
250 mL
1000 mL
5000 mL
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
103-83-3
C9H13N
135.21
MFCD00008329
XXBDWLFCJWSEKW-UHFFFAOYSA-N
n,n-dimethylbenzylamine, benzyldimethylamine, n-benzyldimethylamine, dimethylbenzylamine, bdma, benzenemethanamine, n,n-dimethyl, benzyl-n,n-dimethylamine, n-phenylmethyl dimethylamine, n,n-dimethylbenzenemethanamine, araldite accelerator 062
7681
N,N-dimethyl-1-phenylmethanamine
CN(C)CC1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
103-83-3
C9H13N
135.21
MFCD00008329
XXBDWLFCJWSEKW-UHFFFAOYSA-N
n,n-dimethylbenzylamine, benzyldimethylamine, n-benzyldimethylamine, dimethylbenzylamine, bdma, benzenemethanamine, n,n-dimethyl, benzyl-n,n-dimethylamine, n-phenylmethyl dimethylamine, n,n-dimethylbenzenemethanamine, araldite accelerator 062
7681
N,N-dimethyl-1-phenylmethanamine
CN(C)CC1=CC=CC=C1

N,N-Dimethylbenzylamine is used in the preparation of bis[(N,N-dimethylamino)benzyl] selenide. It acts as a catalyst in the curing reaction of formulations of diglycidyl ether of bisphenol A and tetrahydrophthalic anhydride. It undergoes directed ortho metalation with butyl lithium. It reacts with methyl iodide to get ammonium salt, which is used as phase transfer catalysts. Further, it is used as a catalyst for the formation of polyurethane foams and epoxy resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylbenzylamine is used in the preparation of bis[(N,N-dimethylamino)benzyl] selenide. It acts as a catalyst in the curing reaction of formulations of diglycidyl ether of bisphenol A and tetrahydrophthalic anhydride. It undergoes directed ortho metalation with butyl lithium. It reacts with methyl iodide to get ammonium salt, which is used as phase transfer catalysts. Further, it is used as a catalyst for the formation of polyurethane foams and epoxy resins.

Solubility
Slightly miscible with water.

Notes
Air sensitive. Incompatible with strong acids, strong bases, strong oxidizing agents, strong reducing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY
Melting Point -75°C
Density 0.9
Boiling Point 183°C to 184°C
Flash Point 54°C (129°F)
Linear Formula C6H5CH2N(CH3)2
Refractive Index 1.501
Quantity 250 mL
UN Number UN2619
Beilstein 1099620
Sensitivity Air sensitive
Solubility Information Slightly miscible with water.
Formula Weight 135.21
Percent Purity ≥98%
Chemical Name or Material N-Benzyldimethylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'