missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Benzylbenzamide, 99+%

Brand:  Alfa Aesar™ A12204.22

Code : NEW

Additional Details : CAS Number : 1485-70-7 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11434873

  • £105.26 / 100g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Description

Description

Applications
N-Benzylbenzamide is a convenient precursor to α-substituted benzyl amines and an indicator for the titration of butyl lithium and other lithium bases.

Solubility
Soluble in Acetone (25 mg/ml).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
Specifications

Chemical Identifiers

N-Benzylbenzamide
C6H5CONHCH2C6H5
MFCD00003070
N-Benzoylbenzylamine
1485-70-7
211.26
2211315

Specifications

N-Benzylbenzamide
≥99%
211.26
MFCD00003070
100g
1485-70-7
C6H5CONHCH2C6H5
104° to 108°C
2211315
N-Benzoylbenzylamine
Videos
SDS