missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Benzyl-N,N-bis(2-chloroethyl)amine hydrochloride, 95%

Brand:  Alfa Aesar™ H64057.03

Code : NEW

Additional Details : CAS Number : 10429-82-0 Weight : 0.00100kg Transport : UN number : 2923 Chem class : 8 Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 15438676

  • £41.87 / 1g


Estimated Shipment date
from Supplier 17-07-2018
Add to basket

Specifications

Chemical Identifiers

N-Benzyl-N,N-bis(2-chloroethyl)amine hydrochloride
268.61
UN2923
10429-82-0
MFCD00185654
3916139

Specifications

N-Benzyl-N,N-bis(2-chloroethyl)amine hydrochloride
Powder
268.61
MFCD00185654
3916139
1g
10429-82-0
95%
147° to 150°C
UN2923
White
Videos
SDS